
4-Methyl-1,2,3-thiadiazole-5-carbohydrazide ≥98.0% (by GC, titration analysis)
Catalog # TCIAM3077-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:141.0 to 145.0 deg-C
Specification Test Results
About this item
:
:158.18 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:75423-15-3
Formula:C₄H₆N₄OS
MDL Number:MFCD00052212