Mephenesin ≥98.0% (by GC)
TCIAM3076-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:69.0 to 73.0 deg-C
:
:182.22 g/mol
:154°C
:200-427-4
:GHS07
:59-47-2
:C₁₀H₁₄O₃
:MFCD00004718