Methyl-2-amino-3-chlorobenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:77820-58-7
:
Formula:C₈H₈ClNO₂
MDL Number:MFCD06797833
:185.6100 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 98.0 %
- Melting point:35.0 to 39.0 deg-C
- Acid Value:max. 2.0
Specification Test Results
Frequently Bought Together