1,1'-[Methylenebis(2,6-diethyl-4,1-phenylene)]bis[3-(2-nitrophenyl)urea] ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:2414546-93-1
:
Formula:C₃₅H₃₈N₆O₆
:638.73 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together