
4-(Methylcarbamoyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:121177-82-0
:
Formula:C₈H₁₀BNO₃
MDL Number:MFCD03788427
:178.98 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):96.0 to 111.0 %
Specification Test Results
Frequently Bought Together