
1-Methyl-3-pentylimidazolium bromide ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:343851-31-0
:
Formula:C₉H₁₇BrN₂
MDL Number:MFCD19382537
:233.15 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Brown clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together