Order Entry
Netherlands
ContactUsLinkComponent
5-Methoxy-o-toluidine ≥98.0% (by GC, titration analysis)
Irritant
5-Methoxy-o-toluidine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
5-Methoxy-o-toluidine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    42.0 to 46.0 deg-C

Specification Test Results

Frequently Bought Together