9-Methyl-9H-fluoren-9-ol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6311-22-4
:
Formula:C₁₄H₁₂O
MDL Number:MFCD00183690
:196.25 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:175.0 to 179.0 deg-C
Specification Test Results
Frequently Bought Together