
Methyl-1-methylpiperidine-4-carboxylate ≥98.0% (by GC, titration analysis)
: TCI
:1690-75-1
:
:C₈H₁₅NO₂
:MFCD06658483
:157.21 g/mol
:202°C
:GHS07
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Acid Value:max. 3.0
Frequently Bought Together