Order Entry
Netherlands
ContactUsLinkComponent
Methyl-2-amino-4-(trifluoromethyl)benzoate ≥98.0% (by GC, titration analysis)
Irritant
Methyl-2-amino-4-(trifluoromethyl)benzoate ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Methyl-2-amino-4-(trifluoromethyl)benzoate ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    61.0 to 65.0 deg-C

Specification Test Results

Frequently Bought Together