
Methyl 4-Bromo-2-chlorobenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:185312-82-7
:
Formula:C₈H₆BrClO₂
MDL Number:MFCD10566823
:249.49 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together