
Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate ≥98.0% (by GC)
: TCI
:10250-61-0
:
:C₇H₁₀N₂O₂
:MFCD06661312
:154.17 g/mol
:144°C
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:73.0 to 77.0 deg-C
Frequently Bought Together