6-Methoxyisatin ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:52351-75-4
:
Formula:C₉H₇NO₃
MDL Number:MFCD00667731
:177.1568 g/mol
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:229.0 to 233.0 deg-C
Specification Test Results
Frequently Bought Together