4-(Methoxycarbonyl)bicyclo[2.2.2]octane-1-carboxylic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAM2855-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:186.0 to 190.0 deg-C
Specification Test Results
About this item
Synonyms: Bicyclo[2.2.2]octane-1,4-dicarboxylic acid monomethyl ester, Monomethyl Hydrogen Bicyclo[2.2.2]octane-1,4-dicarboxylate, 4-(Methoxycarbonyl)bicyclo[2.2.2]octan-1-carboxylic acid
:
:212.2423 g/mol
:GHS07
CAS Number:18720-35-9
:MFCD08274673
Formula:C₁₁H₁₆O₄
MDL Number:MFCD18459359