Order Entry
Netherlands
ContactUsLinkComponent
4-Methoxy-2-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Irritant
4-Methoxy-2-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAM2853-1G
Supplier:  TCI
CAS Number:  
Irritant
4-Methoxy-2-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAM2853-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    98.0 to 109.0 %

Specification Test Results

About this item

 :
 :219.95 g/mol
 :GHS07
CAS Number:313546-16-6
Formula:C₈H₈BF₃O₃
MDL Number:MFCD03095346