Order Entry
Netherlands
ContactUsLinkComponent
2-Mercaptopyridine-N-oxide sodium salt, anhydrous ≥98.0% (by HPLC)
Irritant
2-Mercaptopyridine-N-oxide sodium salt, anhydrous ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
Irritant
2-Mercaptopyridine-N-oxide sodium salt, anhydrous ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Iodometric Titration):
    min. 97.0 %

Specification Test Results

Frequently Bought Together