
Methyl 4-Bromo-3-chlorobenzoate ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:117738-74-6
:
Formula:C₈H₆BrClO₂
MDL Number:MFCD11045619
:249.49 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:57.0 to 61.0 deg-C
Specification Test Results
Frequently Bought Together