
2-(Methoxycarbonyl)-6-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:21606-04-2
:
Formula:C₉H₇NO₆
MDL Number:MFCD00034781
:225.16 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:160.0 to 164.0 deg-C
Specification Test Results
Frequently Bought Together