

1-((4-Chlorophenyl)(phenyl)methyl)-4-(3-methylbenzyl)piperazine dihydrochloride ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1104-22-9
:
Formula:C₂₅H₂₉Cl₃N₂
MDL Number:MFCD00058199
:463.87 g/mol
EINECS:214-164-8
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:210.0 to 214.0 deg-C
Specification Test Results
Frequently Bought Together