Order Entry
Netherlands
ContactUsLinkComponent
Methylcobalamin hydrate ≥98.0% (by total nitrogen basis)
Methylcobalamin hydrate ≥98.0% (by total nitrogen basis)
Catalog # TCIAM2742-1G
Supplier:  TCI
CAS Number:  
Methylcobalamin hydrate ≥98.0% (by total nitrogen basis)
Catalog # TCIAM2742-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Red to Dark red to Brown powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 % (calcd.on anh.substance)
  • Water:
    max. 12.0 %

Specification Test Results

About this item

 :
 :1344.41 g/mol (anhy)
Storage Temperature:Refrigerator
EINECS:236-535-3
CAS Number:288315-09-3
 :13422-55-4
Formula:C₆₃H₉₁CoN₁₃O₁₄P·xH₂O
MDL Number:MFCD00082483