
1-(2-Methoxyethyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAM2597-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:74.0 to 78.0 deg-C
Specification Test Results
About this item
:
:134.2000 g/mol
:GHS07
CAS Number:102353-42-4
Formula:C₄H₁₀N₂OS
MDL Number:MFCD00060469