
2-Methoxy-4-nitrobenzonitrile ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:101084-96-2
:
Formula:C₈H₆N₂O₃
MDL Number:MFCD02093781
:178.15 g/mol
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:177.0 to 181.0 deg-C
Specification Test Results
Frequently Bought Together