
6-Methyl-2-(p-tolylamino)-4H-3,1-benzoxazin-4-one ≥95.0% (by HPLC)
: TCI
:86672-58-4
:
:C₁₆H₁₄N₂O₂
:MFCD03272508
:266.30 g/mol
:GHS07
:Freezer
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Elemental analysis (Nitrogen):10.00 to 11.00 %
Frequently Bought Together