[6,6]-Phenyl-C71-butyric acid methyl ester (mixture of isomers) ≥98.0% (by HPLC)
Catalog # TCIAM2550-50MG
Supplier: TCI
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Appearance:Black powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Carbon):88.00 to 96.00 %
Specification Test Results
About this item
:
:1031.01 g/mol
CAS Number:609771-63-3
Formula:C₈₂H₁₄O₂
MDL Number:MFCD09842750