
Methyl-5-amino-2-methylbenzoate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:18595-12-5
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD08752568
:165.19 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:40.0 to 44.0 deg-C
Specification Test Results
Frequently Bought Together