Order Entry
Netherlands
ContactUsLinkComponent
Monomethyl-trans-1,4-cyclohexanedicarboxylate ≥97.0% (by GC, titration analysis)
Irritant
Monomethyl-trans-1,4-cyclohexanedicarboxylate ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Monomethyl-trans-1,4-cyclohexanedicarboxylate ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    126.0 to 130.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together