Order Entry
Netherlands
ContactUsLinkComponent
 
 
Methyl 2,2':6',2''-Terpyridine-4'-carboxylate ≥98.0% (by HPLC)
  TCIAM2464-100MG
 :  TCI
 :  
undefined
 
Methyl 2,2':6',2''-Terpyridine-4'-carboxylate ≥98.0% (by HPLC)
  TCIAM2464-100MG
 :  TCI
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    208.0 to 212.0 deg-C

 

 

 :
 :291.31 g/mol
 :GHS07
 :247058-06-6
 :C₁₇H₁₃N₃O₂
 :MFCD11042546