Order Entry
Netherlands
ContactUsLinkComponent
Meclofenoxate hydrochloride ≥98.0% (by titrimetric analysis)
Meclofenoxate hydrochloride ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Meclofenoxate hydrochloride ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    137.0 to 142.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together