
Methyl-1H-pyrazole-3-carboxylate ≥98.0% (by GC)
Catalog # TCIAM2444-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:144.0 to 148.0 deg-C
Specification Test Results
About this item
:
:126.11 g/mol
:GHS07
CAS Number:15366-34-4
Formula:C₅H₆N₂O₂
MDL Number:MFCD00649381