Methyl-4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAM2426-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:312.33 g/mol
CAS Number:155405-80-4
Formula:C₁₆H₁₆N₄O₃
MDL Number:MFCD17976681