(6R,7R)-4-Methoxybenzyl-3-(chloromethyl)-8-oxo-7-(2-phenylacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:104146-10-3
:
Formula:C₂₄H₂₃ClN₂O₅S
MDL Number:MFCD00191253
:486.967 g/mol
Storage Temperature:Freezer
EINECS:403-040-0
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together