
2-Methoxy-4-nitrobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
: TCI
:21320-91-2
:
:C₇H₆ClNO₅S
:MFCD03094697
:251.65 g/mol
:3261
:8,III
:GHS05
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:97.0 to 101.0 deg-C
Frequently Bought Together