


Methylene blue solution in methanol for cell staining
Catalogus # TCIAM2392-100ML
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Blue to Dark blue clear liquid to cloudy liquid
- Absorbance:min. 350 (664 nm)
- Cell staining test:Extent of staining same as reference
Specification Test Results
About this item
:
:319.86 g/mol
Merck Index:14,06060
EINECS:200-515-2
UN:1230
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:61-73-4
:MFCD00012111
Formula:C₁₆H₁₈ClN₃S
MDL Number:MFCD00150008