Order Entry
Netherlands
ContactUsLinkComponent
Methyl-3,4-diaminobenzoate ≥98.0% (by GC, titration analysis)
Irritant
Methyl-3,4-diaminobenzoate ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Methyl-3,4-diaminobenzoate ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Gray to Red powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    107.0 to 111.0 deg-C

Specification Test Results

Frequently Bought Together