Order Entry
Netherlands
ContactUsLinkComponent
Methylamine ∼9% ∼2 mol/L in isopropanol (IPA)
Flammable
Corrosive
Irritant
Health
Methylamine ∼9% ∼2 mol/L in isopropanol (IPA)
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Methylamine ∼9% ∼2 mol/L in isopropanol (IPA)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Neutralization titration):
    8.5 to 11.0 w/w%

Specification Test Results

Frequently Bought Together