

Minocycline hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13614-98-7
:
Formula:C₂₃H₂₇N₃O₇
MDL Number:MFCD00083669
:493.94 g/mol
Merck Index:14,06202
EINECS:237-099-7
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:-170 to -180 deg (C=0.5、0.1mol/L HCl)
- Water:4.3 to 8.0 %
Specification Test Results
Frequently Bought Together