Order Entry
Netherlands
ContactUsLinkComponent
 
 
6-Mercapto-1-hexanol ≥98.0% (by GC)
 :  TCI
 
undefined
 
6-Mercapto-1-hexanol ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

Frequently Bought Together