Methyl-1H-imidazole-5-carboxylate ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:17325-26-7
:
Formule:C₅H₆N₂O₂
MDL-nummer:MFCD00216584
:126.11 g/mol
Specificatie testresultaten
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:151.0 to 156.0 deg-C
Specificatie testresultaten
Frequently Bought Together