Order Entry
Netherlands
ContactUsLinkComponent
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ∼18% ∼1.0 mol/L in dichloromethane for determination of the optical purity of alcohols and amines
Corrosive
Irritant
Health
Environment
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ∼18% ∼1.0 mol/L in dichloromethane for determination of the optical purity of alcohols and amines
Catalog # TCIAM2215-5G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Irritant
Health
Environment
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ∼18% ∼1.0 mol/L in dichloromethane for determination of the optical purity of alcohols and amines
Catalog # TCIAM2215-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Optical purity (GC):
    min. 99.0 ee%
  • Concentration (Argentmetric titration):
    18.0 to 20.0 w/w%

Specification Test Results

About this item

 :
 :252.62 g/mol
Merck Index:14,06280
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
 :GHS05
 :GHS07
 :GHS08
 :GHS09
CAS Number:39637-99-5
Formula:C₁₀H₈ClF₃O₂
MDL Number:MFCD00044400