
Methyl-3-hydroxy-4-methoxybenzoate ≥98.0% (by GC)
: TCI
:6702-50-7
:
:C₉H₁₀O₄
:MFCD01321262
:182.1733 g/mol
:GHS07
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:62.0 to 66.0 deg-C
Frequently Bought Together