
Methyl-2-bromo-2-(2-chlorophenyl)acetate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:85259-19-4
:
Formula:C₉H₈BrClO₂
MDL Number:MFCD08444353
:263.5156 g/mol
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together