Order Entry
Netherlands
ContactUsLinkComponent
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol ≥98.0% (by GC, titration analysis)
Irritant
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    72.0 to 76.0 deg-C
  • Specific rotation [a]20/D:
    -10.0 to -14.0 deg (C=1, methanol)

Specification Test Results

Related Information

Frequently Bought Together