
2-Methylpyrrolidine hydrochloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAM2124-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
About this item
:
:121.61 g/mol
:GHS07
CAS Number:54677-53-1
Formula:C₅H₁₁N
MDL Number:MFCD08274961