

3-Methylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
: TCI
:6850-35-7
:
:C₇H₁₅N
:MFCD00001494
:113.20 g/mol
:151°C
:0.85
:229-940-1
:2733
:3,II
:GHS02
:GHS05
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomer)
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together