Order Entry
Netherlands
ContactUsLinkComponent
 
1-Methyl-1-[4-(diphenylphosphino)benzyl]pyrrolidinium bromide ≥98.0% (by HPLC)
Catalog # TCIAM2103-1G
Supplier:  TCI
CAS Number:  
1-Methyl-1-[4-(diphenylphosphino)benzyl]pyrrolidinium bromide ≥98.0% (by HPLC)
Catalog # TCIAM2103-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Iodometric Titration):
    min. 97.0 %

Specification Test Results

About this item

 :
 :440.36 g/mol
CAS Number:1229444-44-3
Formula:C₂₄H₂₇BrNP