[6,6]-Phenyl-C61-butyric acid methyl ester ≥99.5% (by HPLC)
Catalog # TCIAM2088-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Dark green to Dark red to Black powder to crystal
- Purity (HPLC):min. 99.5 area%
Specification Test Results
About this item
:
:910.90 g/mol
CAS Number:160848-22-6
Formula:C₇₂H₁₄O₂
MDL Number:MFCD07784544