Order Entry
Netherlands
ContactUsLinkComponent
Methyl-3-bromo-2-(bromomethyl)propionate ≥97.0% (by GC)
Irritant
Methyl-3-bromo-2-(bromomethyl)propionate ≥97.0% (by GC)
Supplier:  TCI
Certificates
Irritant
Methyl-3-bromo-2-(bromomethyl)propionate ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light red to Green clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

Frequently Bought Together