Order Entry
Netherlands
ContactUsLinkComponent
2-Methyl-4-propyl-1,3-oxathiane (cis- and trans- mixture) cis- and trans-mixture ≥97.0% (by GC)
2-Methyl-4-propyl-1,3-oxathiane (cis- and trans- mixture) cis- and trans-mixture ≥97.0% (by GC)
Catalog # TCIAM2049-5G
Supplier:  TCI
CAS Number:  
2-Methyl-4-propyl-1,3-oxathiane (cis- and trans- mixture) cis- and trans-mixture ≥97.0% (by GC)
Catalog # TCIAM2049-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white clear liquid
  • Purity (GC):
    min. 97.0 % (total of isomer)

Specification Test Results

About this item

Synonyms: 2-Methyl-4-propyl-1,3-oxathiane
 :
Flash Pt:50°C
UN:1993
ADR:3,III
 :160.28 g/mol
 :GHS02
CAS Number:67715-80-4
Formula:C₈H₁₆OS
MDL Number:MFCD00673203