Methyl-5-(ethylsulphonyl)-2-methoxybenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:62140-67-4
:
Formula:C₁₁H₁₄O₅S
MDL Number:MFCD01317547
:258.29 g/mol
EINECS:263-429-4
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:124.0 to 128.0 deg-C
Specification Test Results
Frequently Bought Together