Methyl-2-methoxy-5-(methylsulphonyl)benzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:63484-12-8
:
Formula:C₁₀H₁₂O₅S
MDL Number:MFCD01317545
:244.26 g/mol
EINECS:264-267-7
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:115.0 to 119.0 deg-C
Specification Test Results
Frequently Bought Together